missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ ERBB2 Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Thr 652) of human ERBB2 (NP_004439.2) fused to the Fc region of human IgG1 at the C-terminus

$89.50 - $1,384.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias ErbB2, NEU, NGL, HER2, TKR1, CD340, HER-2, c-erb B2, HER-2/neu
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10004H02H25 Life Technologies
LS10004H02H25
5 x 5μg Each for $225.00
10004H02H5 Life Technologies
LS10004H02H5
5μg Each for $89.50
10004H02H50 Life Technologies
LS10004H02H50
50μg Each for $390.00
10004H02HL Life Technologies
LS10004H02H250
5 x 50μg Each for $1,384.00
Specifications

Description

V-erb-b2 erythroblastic leukemia viral oncogene homolog 2 (ERBB2), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human ERBB2 at 2.5μg/mL can bind Herceptin with a linear range of 3.2-80ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥85% by SDS-PAGE
<1 EU/μg
Lyophilized
ErbB2, NEU, NGL, HER2, TKR1, CD340, HER-2, c-erb B2, HER-2/neu