Please login to your online account to display your discounted pricing

Sino Biological™ EPOR Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 250) of human EPOR precursor (NP_000112.1) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10707H08H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10707H08H5

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 26.3kDa
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Gene Alias EPOR, mgC138358
Structural Form Truncated/Recombinant
Protein Subtype Cytokine Receptors
Protein Family Hormones & Receptors
Common Name Erythropoietin Receptor
Gene ID (Entrez) 2057
Quantity 5μg
Endotoxin Concentration <1 EU/μg
Protein Tag His Tag
Validated Application Assay Standard
Purity ≥90% by SDS-PAGE
Expression System Mammalian
Gene Symbol EPOR
Format Lyophilized

Erythropoietin receptor (EPOR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CSF2RB-His at 1μg/well can bind biotinylated human EPOR-His with a linear range of 0.16-4.0μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.