missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ EPOR & CSF2RB (CD131) Recombinant Human Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 250) of human EPOR (NP_000112.1) linked by a peptide to the extracellular domain (Trp 17-Trp 443) of human CD131 (NP_000386.1) and fused to the Fc region of human IgG1 at the C-terminus

$440.00 - $440.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD131, EPOR
Gene ID (Entrez) 2057/1439
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CT010H02H50 Life Technologies
LSCT010H02H50
50μg Each for $440.00
Specifications

Description

Erythropoietin receptor & colony stimulating factor 2 receptor, beta, low-affinity (EPOR & CSF2RB/ CD131), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥85% by SDS-PAGE
Lyophilized
2057/1439
<1 EU/μg
CD131, EPOR