Please login to your online account to display your discounted pricing

Sino Biological™ EPOR & CSF2RB (CD131) Recombinant Human Protein, hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 250) of human EPOR (NP_000112.1) linked by a peptide to the extracellular domain (Trp 17-Trp 443) of human CD131 (NP_000386.1) and fused to the Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LSCT010H02H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. CT010H02H50

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 2057/1439
Molecular Weight 125kDa
Purity ≥85% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Truncated/Recombinant/Cell Surface Protein
Protein Subtype Cytokine Receptors
Endotoxin Concentration <1 EU/μg
Gene Alias CD131, EPOR
Common Name Erythropoietin Receptor/Colony stimulating factor 2 receptor, beta, low-affinity
Gene Symbol EPOR CSF2RB
Validated Application Assay Standard
Protein Family CD Proteins
Expression System Mammalian
Protein Tag IgG1 Fc Tag

Erythropoietin receptor & colony stimulating factor 2 receptor, beta, low-affinity (EPOR & CSF2RB/ CD131), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.