missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ EPHA4 Recombinant Human Protein (without Catalytic Activity Domain), His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human EPHA4 (NP_031962.2) extracellular domain (Met 1-Thr 547) was expressed, with a polyhistidine tag at the C-terminus

$365.00 - $365.00

Specifications

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11314H08H50 Life Technologies
LS11314H08H50
50μg Each for $365.00
Specifications

Description

Eph Receptor A4 (EPHA4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its ability to bind biotinylated Human EphrinA5-his (Mfr. No. 10192-H08H) in functional ELISA
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

0.0