missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EPCAM Recombinant Human Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Lys265) of human EPCAM (NP_002345.1) fused to Fc region of human IgG1 at the C-terminus

$223.00 - $223.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias TACSTD1, EGP, KSA, M4S1, MK-1, CD326, EGP40, MIC18, TROP1, Ep-CAM, hEGP-2, CO17-1A
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10694H02H25 View Documents Life Technologies
LS10694H02H25
5 x 5μg Each for $223.00
 
Description

Description

Epithelial Cell Adhesion Molecule (EPCAM), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, 100mM Glycine, 10mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
TACSTD1, EGP, KSA, M4S1, MK-1, CD326, EGP40, MIC18, TROP1, Ep-CAM, hEGP-2, CO17-1A
PAGE
Lyophilized
4072

For Research Use Only. Not for use in diagnostic procedures.