missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ ENO1 Recombinant Human Protein, N-His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human ENO1 (P06733-1) (Met 1-Lys 434) fused to a polyhistidine tag at the N-terminus

$93.00 - $782.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Format Frozen
Gene Alias ENO1L1, MPB1, NNE, PPH
Gene ID (Entrez) 2023
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11554H07E25 Life Technologies
LS11554H07E25
5 x 5μg Each for $191.00
11554H07E5 Life Technologies
LS11554H07E5
5μg Each for $93.00
11554H07E50 Life Technologies
LS11554H07E50
50μg Each for $269.00
11554H07EL Life Technologies
LS11554H07E250
5 x 50μg Each for $782.00
Specifications

Description

Enolase 1 alpha (ENO1), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM Tris 10% glycerol, pH 8.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Frozen
2023
PAGE
ENO1L1, MPB1, NNE, PPH