Please login to your online account to display your discounted pricing

Sino Biological™ ENO1 Recombinant Human Protein, N-His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human ENO1 (P06733-1) (Met 1-Lys 434) fused to a polyhistidine tag at the N-terminus

Manufacturer: life technologies  LS11554H07E5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11554H07E5

  • / Each
 

Description & Specifications

Specifications

Quantity 5μg
Common Name Enolase 1 Alpha
Gene Symbol ENO1
Molecular Weight 49.3kDa
Purity ≥90% by SDS-PAGE
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Frozen
Gene Alias ENO1L1, MPB1, NNE, PPH
Validated Application Assay Standard
Expression System E. coli
Protein Tag N-His
Gene ID (Entrez) 2023
Purification Method PAGE
Structural Form Full Length/Recombinant
Protein Family Other Proteins
Storage Requirements Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.

Enolase 1 alpha (ENO1), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM Tris 10% glycerol, pH 8.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.