Please login to your online account to display your discounted pricing

Sino Biological™ ENG Recombinant Human Protein , hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 586) of human ENG (NP_001108225.1) fused to the human IgG1 Fc region at the C-terminus

Manufacturer: life technologies  LS10149H02H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10149H02H5

  • / Each
 

Description & Specifications

Specifications

Quantity 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype TGFs (Transforming Growth Factors)
Validated Application Assay Standard
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Molecular Weight 87.5kDa
Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Gene Alias RP11-228B15.2, CD105, END, FLJ41744, HHT1, ORW, ORW1
Common Name Endoglin
Gene Symbol ENG
Protein Tag IgG1 Fc Tag
Protein Family CD Proteins
Expression System Mammalian
Format Lyophilized

Endoglin (ENG), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to bind biotinylated human TGFRB1 in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.