missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EIF4EBP1 Recombinant Human Protein, N-His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human 4EBP1 (NP_004086.1) (Ser 2-Ile 118) fused to a polyhistidine tag at the N-terminus

$200.00 - $864.00

Specifications

Purity ≥85% by SDS-PAGE
Purification Method PAGE
Format Frozen
Gene Alias 4E-BP1, EIF4EBP1, BP-1, mgC4316, PHAS-I
Gene ID (Entrez) 1978
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10022H07E25 View Documents Life Technologies
LS10022H07E25
5 x 5μg Each for $200.00
 
10022H07EL View Documents Life Technologies
LS10022H07E250
5 x 50μg Each for $864.00
 
Description

Description

Eukaryotic Translation Initiation Factor 4E Binding Protein 1 (EIF4EBP1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM Tris, 500mM NaCl, 10% glycerol, pH 8.0
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
Frozen
1978
PAGE
4E-BP1, EIF4EBP1, BP-1, mgC4316, PHAS-I

For Research Use Only. Not for use in diagnostic procedures.