Please login to your online account to display your discounted pricing

Sino Biological™ EIF4EBP1 Recombinant Human Protein, N-His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human 4EBP1 (NP_004086.1) (Ser 2-Ile 118) fused to a polyhistidine tag at the N-terminus

Manufacturer: life technologies  LS10022H07E25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10022H07E25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 13.4kDa
Purification Method PAGE
Purity ≥85% by SDS-PAGE
Quantity 5 x 5μg
Storage Requirements Store at -5° to -30°C. Store aliquots under sterile conditions at -5° to -30°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Full Length/Recombinant
Gene Alias 4E-BP1, EIF4EBP1, BP-1, mgC4316, PHAS-I
Common Name Eukaryotic Translation Initiation Factor 4E Binding Protein 1
Gene Symbol EIF4EBP1
Protein Tag N-His
Validated Application Assay Standard
Protein Family Other Proteins
Expression System E. coli
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Frozen
Gene ID (Entrez) 1978

Eukaryotic Translation Initiation Factor 4E Binding Protein 1 (EIF4EBP1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM Tris, 500mM NaCl, 10% glycerol, pH 8.0
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.