missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EGFR Recombinant Mouse Protein (without Catalytic Activity Domain), hIgG1-Fc Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse EGFR (Q01279) (Met 1-Ser 647) fused to the Fc region of human IgG1 at the C-terminus

$96.50 - $96.50

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias RP23-295E4.1, 9030024J15Rik, AI552599, Erbb, Errp, Wa5, wa-2, wa2
View More Specs

Includes: Five vials of lyophilized protein

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
51091M02H5 View Documents Life Technologies
LS51091M02H5
5μg Each for $96.50
 
Description

Description

Epidermal Growth Factor Receptor (EGFR), recombinant mouse protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its ability to bind human EGF in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
<1 EU/μg
Lyophilized
RP23-295E4.1, 9030024J15Rik, AI552599, Erbb, Errp, Wa5, wa-2, wa2

For Research Use Only. Not for use in diagnostic procedures.