missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EGFR Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 645) of human EGFR (NP_005219) fused to the Fc region of human IgG1 at the C-terminus

$95.75 - $413.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias EGF R, ErbB, HER1, mENA, ErbB1, PIG61
Gene ID (Entrez) 1956
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10001H02H5 View Documents Life Technologies
LS10001H02H5
5μg Each for $95.75
 
10001H02H50 View Documents Life Technologies
LS10001H02H50
50μg Each for $413.00
 
Description

Description

Epidermal growth factor receptor (EGFR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human EGF at 10μg/mL (100μL/well) can bind human EGFR with a linear range of 0.64-400ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
Lyophilized
1956
<1 EU/μg
EGF R, ErbB, HER1, mENA, ErbB1, PIG61

For Research Use Only. Not for use in diagnostic procedures.