Please login to your online account to display your discounted pricing

Sino Biological™ EGFR Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 645) of human EGFR (NP_005219) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS10001H02H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10001H02H50

  • / Each
 

Description & Specifications

Specifications

Format Lyophilized
Common Name Epidermal growth factor receptor
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Expression System Mammalian
Gene ID (Entrez) 1956
Protein Subtype Receptor Tyrosine Kinases|Growth Factor Receptors
Protein Family Growth Factors & Receptors
Gene Alias EGF R, ErbB, HER1, mENA, ErbB1, PIG61
Molecular Weight 95kDa
Structural Form Truncated/Recombinant
Protein Tag IgG1 Fc Tag
Quantity 50μg
Purity ≥85% by SDS-PAGE
Gene Symbol EGFR
KinaseFamily EGF Receptor (ErbB) Kinase Family
Endotoxin Concentration <1 EU/μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Validated Application Assay Standard

Epidermal growth factor receptor (EGFR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human EGF at 10μg/mL (100μL/well) can bind human EGFR with a linear range of 0.64-400ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.