Please login to your online account to display your discounted pricing

Sino Biological™ EGF Recombinant Mouse Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of mouse EGF (P01132) (Asn 977-Arg 1029) fused to the Fc region of human IgG1 at the N-terminus

$77.50 - $226.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias AI790464
View More Specs
Includes:  Five vials of lyophilized protein

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50482M01H25

DFS Item
 
life technologies
LS50482M01H25
5 x 5μg Each for $92.75

50482M01H5

DFS Item
 
life technologies
LS50482M01H5
5μg Each for $77.50

50482M01H50

DFS Item
 
life technologies
LS50482M01H50
50μg Each for $124.00

50482M01HL

DFS Item
 
life technologies
LS50482M01H250
5 x 50μg Each for $226.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias AI790464

Epidermal growth factor (EGF), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard, or in other research applications.

  • Activity: Measured in a cell proliferation assay using BALB/c 3T3 mouse embryonic fibroblasts; the EC50 for this effect is typically 0.2-1ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.