Please login to your online account to display your discounted pricing

Sino Biological™ EGF Recombinant Human Protein, hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the mature soluble form of human EGF (NP_001954.2) (Asn 971-Arg 1023) fused to the Fc region of human IgG1 at the N-terminus

Manufacturer: life technologies  LS10605H01H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10605H01H50

  • / Each
 

Description & Specifications

Specifications

Protein Subtype EGF (Epidermal Growth Factor)
Endotoxin Concentration <1 EU/μg
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Molecular Weight 33kDa
Purity ≥85% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Full Length/Recombinant
Gene Alias HOMG4, URG, EGF
Common Name Epidermal growth factor
Gene Symbol EGF
Protein Tag IgG1 Fc Tag
Validated Application Assay Standard
Protein Family Growth Factors & Receptors
Format Lyophilized

Epidermal growth factor (EGF), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard, or in other research applications.

  • Activity: Measured in a cell proliferation assay using BALB/c 3T3 mouse embryonic fibroblasts, the EC50 for this effect is typically 20-80pg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.