missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EGF Recombinant Canine Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form (Asn 973-Arg 1024) of canine EGF (Q9BEA0) fused to a polyhistidine tag at the N-terminus

$126.00 - $389.00

Specifications

Purity ≥95% by SDS-PAGE
Format Lyophilized
Gene Alias CEGF
Gene ID (Entrez) 403657
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
70013D07E5 View Documents Life Technologies
LS70013D07E5
5μg Each for $126.00
 
70013D07E50 View Documents Life Technologies
LS70013D07E50
50μg Each for $389.00
 
Description

Description

Epidermal growth factor (EGF), recombinant canine protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: Measured in a cell proliferation assay using BALB/c 3T3 mouse embryonic fibroblasts; the ED50 for this effect is typically 0.1-0.5ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥95% by SDS-PAGE
Lyophilized
CEGF
403657

For Research Use Only. Not for use in diagnostic procedures.