missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EFNB2 (EphrinB2) Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 232) of mouse EFNB2 (NP_034241.2) fused to a polyhistidine tag at the C-terminus

$202.00 - $202.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias EFNB2, ELF-2, Epl5, Eplg5, Htk-L, LERK-5, Lerk5, NLERK-1
Gene ID (Entrez) 13642
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50598M08H50 View Documents Life Technologies
LS50598M08H50
50μg Each for $202.00
 
Description

Description

Ephrin B2 (EFNB2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to compete with mouse EFNB2-Fc for binding to immobilized mouse EPHB4 in a functional ELISA assay
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
13642
<1 EU/μg
EFNB2, ELF-2, Epl5, Eplg5, Htk-L, LERK-5, Lerk5, NLERK-1

For Research Use Only. Not for use in diagnostic procedures.