Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
missing translation for 'pleaseLogIn'

Sino Biological™ EFNB2 (EphrinB2) Recombinant Mouse Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 232) of mouse EFNB2 (NP_034241.2) fused to the Fc region of human IgG1 at the C-terminus

$80.50 - $80.50

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias EFNB2, ELF-2, Epl5, Eplg5, Htk-L, LERK-5, Lerk5, NLERK-1
Gene ID (Entrez) 13642
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
50598M02H5 Life Technologies
LS50598M02H5
5μg Each for $80.50
Specifications

Description

Ephrin B2 (EFNB2), recombinant mouse protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse EPHB4 at 2μg/mL (100μL/well) can bind mouse EFNB2-Fc with a linear range of 0.2-25ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
13642
<1 EU/μg
EFNB2, ELF-2, Epl5, Eplg5, Htk-L, LERK-5, Lerk5, NLERK-1