missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EFNB2 (EphrinB2) Recombinant Human Protein, hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 229) of human EFNB2 (NP_004084.1) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

$122.00 - $233.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias EFNB2, RP11-272L14.1, EPLG5, HTKL, Htk-L, LERK5, mgC126226, mgC126227, mgC126228
Gene ID (Entrez) 1948
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10881H03H25 View Documents Life Technologies
LS10881H03H25
5 x 5μg Each for $233.00
 
10881H03H5 View Documents Life Technologies
LS10881H03H5
5μg Each for $122.00
 
Description

Description

Ephrin-B2 (EFNB2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human EFNB4-His at 2μg/mL (100μL/well) can bind human EFNB2-Fc with a linear range of 1-25ng/mL, immobilized human EFNB4 at 2μg/mL (100μL/well) can bind human EFNB2-Fc with a linear range of 1-125ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
1948
<1 EU/μg
EFNB2, RP11-272L14.1, EPLG5, HTKL, Htk-L, LERK5, mgC126226, mgC126227, mgC126228

For Research Use Only. Not for use in diagnostic procedures.