missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ EFNB2 (EphrinB2) Recombinant Human Protein, hIgG1-Fc.His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 229) of human EFNB2 (NP_004084.1) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Manufacturer:  Life Technologies LS10881H03H25

 View more versions of this product

Catalog No. 10881H03H25


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; EFNB2 (EphrinB2) Recombinant Human Protein, hIgG1-Fc.His Tag; 5 x 5ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

Ephrin-B2 (EFNB2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human EFNB4-His at 2μg/mL (100μL/well) can bind human EFNB2-Fc with a linear range of 1-25ng/mL, immobilized human EFNB4 at 2μg/mL (100μL/well) can bind human EFNB2-Fc with a linear range of 1-125ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
1948
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Ephrins
EFNB2, RP11-272L14.1, EPLG5, HTKL, Htk-L, LERK5, mgC126226, mgC126227, mgC126228
EFNB2
Kinases & Inhibitors
His Tag/IgG1 Fc Tag
Lyophilized
50.3kDa
5 x 5μg
Truncated/Recombinant/Cell Surface Protein/Ligand
<1 EU/μg
Ephrin-B2
Assay Standard
Mammalian