Please login to your online account to display your discounted pricing

Sino Biological™ EFNB1 (EphrinB1) Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 232) of human EFNB1 (NP_004420.1) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10894H08H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10894H08H50

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 1947
Quantity 50μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Alias EFNB1, RP3-496G1.1, CFND, CFNS, EFL3, EPLG2, Elk-L, LERK2
Validated Application Assay Standard
Endotoxin Concentration <1 EU/μg
Common Name Ephrin-B1
Structural Form Truncated/Recombinant/Cell Surface Protein/Ligand
Expression System Mammalian
Protein Subtype Ephrins
Gene Symbol EFNB1
Protein Family Kinases & Inhibitors
Protein Tag His Tag
Purity ≥90% by SDS-PAGE
Molecular Weight 24.5kDa

Ephrin-B1 (EFNB1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human EFNB1 at 10μg/mL (100μL/well) can bind human EPHB6 with a linear range of 0.16-4μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.