missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ EFNA5 (EphrinA5) Recombinant Mouse Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form (Met 1-Asn 203) of mouse EFNA5 long isoform (O08543-1) fused to a polyhistidine tag at the C-terminus

$93.25 - $396.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias AL-1, AV158822, EFL-5, Ephrin-A5, Epl7, LERK-7, RAGS
Gene ID (Entrez) 13640
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50597M08H25 Life Technologies
LS50597M08H25
5 x 5μg Each for $232.00
50597M08H5 Life Technologies
LS50597M08H5
5μg Each for $93.25
50597M08H50 Life Technologies
LS50597M08H50
50μg Each for $302.00
50597M08HL Life Technologies
LS50597M08H250
5 x 50μg Each for $396.00
Specifications

Description

Ephrin A5 (EFNA5), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse EPHA5 at 2μg/mL (100μL/well) can bind mouse EFNA4 with a linear range of 1.28-32ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
13640
<1 EU/μg
AL-1, AV158822, EFL-5, Ephrin-A5, Epl7, LERK-7, RAGS