missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ EFNA5 (EphrinA5) Recombinant Human Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding (Met 1-Asn 203) of human EphrinA5 precursor (NM_001962.1) fused to the Fc region of human IgG1 at the C-terminus

$80.50 - $380.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias EFNA5, AF1, EFL5, RAGS, EPLG7, GLC1M, LERK7
Gene ID (Entrez) 1946
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10192H02H25 Life Technologies
LS10192H02H25
5 x 5μg Each for $159.00
10192H02H5 Life Technologies
LS10192H02H5
5μg Each for $80.50
10192H02H50 Life Technologies
LS10192H02H50
50μg Each for $210.00
10192H02HL Life Technologies
LS10192H02H250
5 x 50μg Each for $380.00
Specifications

Description

Ephrin-A5 (EFNA5), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human EFNA5-Fc at 1μg/well can bind biotinylated recombinant human EPHA4 with a linear range of 4.0-20μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
1946
<1 EU/μg
EFNA5, AF1, EFL5, RAGS, EPLG7, GLC1M, LERK7