Please login to your online account to display your discounted pricing

Sino Biological™ EFNA5 (EphrinA5) Recombinant Human Protein, hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding (Met 1-Asn 203) of human EphrinA5 precursor (NM_001962.1) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS10192H02H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10192H02H25

  • / Each
 

Description & Specifications

Specifications

Common Name Ephrin-A5
Format Lyophilized
Gene ID (Entrez) 1946
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Symbol EFNA5
Expression System Mammalian
Validated Application Assay Standard
Protein Subtype Ephrins
Molecular Weight 47.9kDa
Endotoxin Concentration <1 EU/μg
Gene Alias EFNA5, AF1, EFL5, RAGS, EPLG7, GLC1M, LERK7
Purity ≥90% by SDS-PAGE
Structural Form Recombinant/Cell Surface Protein/Ligand
Protein Family Kinases & Inhibitors
Protein Tag IgG1 Fc Tag
Quantity 5 x 5μg

Ephrin-A5 (EFNA5), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human EFNA5-Fc at 1μg/well can bind biotinylated recombinant human EPHA4 with a linear range of 4.0-20μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.