Please login to your online account to display your discounted pricing

Sino Biological™ EFNA5 (EphrinA5) Recombinant Human Protein, hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding (Met 1-Asn 203) of human EphrinA5 precursor (NM_001962.1) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS10192H02H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10192H02H5

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 47.9kDa
Protein Tag IgG1 Fc Tag
Gene Alias EFNA5, AF1, EFL5, RAGS, EPLG7, GLC1M, LERK7
Gene Symbol EFNA5
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Quantity 5μg
Protein Family Kinases & Inhibitors
Protein Subtype Ephrins
Format Lyophilized
Endotoxin Concentration <1 EU/μg
Expression System Mammalian
Gene ID (Entrez) 1946
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Purity ≥90% by SDS-PAGE
Validated Application Assay Standard
Common Name Ephrin-A5
Structural Form Recombinant/Cell Surface Protein/Ligand

Ephrin-A5 (EFNA5), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human EFNA5-Fc at 1μg/well can bind biotinylated recombinant human EPHA4 with a linear range of 4.0-20μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.