missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ EFNA4 (EphrinA4) Recombinant Mouse Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the (Met 1-Gly 176) of mouse EFNA4 (NP_031936.2) fused to the Fc region of human IgG1 at the C-terminus

$208.00 - $208.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias AW494418, EFL-2, Ehk1-L, Epl3, LERK-3, mgC129300, mgC129301
Gene ID (Entrez) 13639
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50595M02H50 Life Technologies
LS50595M02H50
50μg Each for $208.00
Specifications

Description

Ephrin A4 (EFNA4), recombinant mouse protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human EPHA7 at 2μg/mL (100μL/well) can bind mouse EFNA4-Fc with a linear range of 0.08-10ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
13639
<1 EU/μg
AW494418, EFL-2, Ehk1-L, Epl3, LERK-3, mgC129300, mgC129301