missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ EFNA3 (EphrinA3) Recombinant Human Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human EphrinA3 (NP_004943.1), residues Met 1 to Ser 213, fused to the Fc region of human IgG1 at the C-terminus

$92.75 - $404.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias EFNA3, EFL2, EPLG3, Ehk1-L, LERK3
Gene ID (Entrez) 1944
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10188H02H25 Life Technologies
LS10188H02H25
5 x 5μg Each for $232.00
10188H02H5 Life Technologies
LS10188H02H5
5μg Each for $92.75
10188H02H50 Life Technologies
LS10188H02H50
50μg Each for $301.00
10188H02HL Life Technologies
LS10188H02H250
5 x 50μg Each for $404.00
Specifications

Description

Ephrin-A3 (EFNA3), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse EPHA6 at 1μg/mL (100μL/well) can bind human EFNA3-Fc, the ED50 of human EFNA3 is 299.2ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
1944
<1 EU/μg
EFNA3, EFL2, EPLG3, Ehk1-L, LERK3