Please login to your online account to display your discounted pricing

Sino Biological™ EFNA3 (EphrinA3) Recombinant Human Protein, hIgG1-Fc.His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the mature form of human EphrinA3 (NP_004943.1), residues 1 to 213, fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS10188H03H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10188H03H5

  • / Each
 

Description & Specifications

Specifications

Quantity 5μg
Protein Tag His Tag/IgG1 Fc Tag
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene ID (Entrez) 1944
Purity ≥90% by SDS-PAGE
Format Lyophilized
Common Name Ephrin-A3
Structural Form Recombinant/Cell Surface Protein/Ligand
Expression System Mammalian
Endotoxin Concentration <1 EU/μg
Protein Subtype Ephrins
Molecular Weight 49.5kDa
Gene Alias EFNA3, EFL2, EPLG3, Ehk1-L, LERK3
Gene Symbol EFNA3
Protein Family Kinases & Inhibitors
Validated Application Assay Standard
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories

Ephrin-A3 (EFNA3), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.