Please login to your online account to display your discounted pricing

Sino Biological™ EFNA3 (EphrinA3) Recombinant Human Protein, DDDDK Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the mature form of human EphrinA3 (NP_004943.1), residues 1 to 213, fused to a DDDDK tag at the C-terminus

Manufacturer: life technologies  LS10188HCCH50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10188HCCH50

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 22kDa
Purity ≥85% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Recombinant/Cell Surface Protein/Ligand
Protein Subtype Ephrins
Endotoxin Concentration <1 EU/μg
Gene Alias EFNA3, EFL2, EPLG3, Ehk1-L, LERK3
Common Name Ephrin-A3
Gene Symbol EFNA3
Protein Tag DDDDK
Protein Family Kinases & Inhibitors
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 1944
Validated Application Assay Standard

Ephrin-A3 (EFNA3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.