missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EFNA1 (EphrinA1) Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human EFNA1 (NP_004419.2), residues 1 to 182, fused to a polyhistidine tag at the C-terminus

$126.00 - $252.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias EFNA1, B61, ECKLG, EFL1, EPLG1, LERK1, TNFAIP4
Gene ID (Entrez) 1942
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10882H08H25 View Documents Life Technologies
LS10882H08H25
5 x 5μg Each for $252.00
 
10882H08H5 View Documents Life Technologies
LS10882H08H5
5μg Each for $126.00
 
Description

Description

Ephrin-A1 (EFNA1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to compete with human EFNA1 for binding to immobilized human EFNA2 in a functional ELISA assay
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
1942
<1 EU/μg
EFNA1, B61, ECKLG, EFL1, EPLG1, LERK1, TNFAIP4

For Research Use Only. Not for use in diagnostic procedures.