Please login to your online account to display your discounted pricing

Sino Biological™ EFNA1 (EphrinA1) Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the mature form of human EFNA1 (NP_004419.2), residues 1 to 182, fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10882H08H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10882H08H5

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 20.8kDa
Purity ≥90% by SDS-PAGE
Quantity 5μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Recombinant/Cell Surface Protein/Ligand
Protein Subtype Ephrins
Endotoxin Concentration <1 EU/μg
Gene Alias EFNA1, B61, ECKLG, EFL1, EPLG1, LERK1, TNFAIP4
Common Name Ephrin-A1
Gene Symbol EFNA1
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Kinases & Inhibitors
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 1942

Ephrin-A1 (EFNA1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to compete with human EFNA1 for binding to immobilized human EFNA2 in a functional ELISA assay
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.