missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EFNA1 (EphrinA1) Recombinant Human Protein, hIgG1-Fc.His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human EFNA1 (NP_004419.2), residues 1 to 182, fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

$126.00 - $249.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias EFNA1, B61, ECKLG, EFL1, EPLG1, LERK1, TNFAIP4
Gene ID (Entrez) 1942
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10882H03H25 View Documents Life Technologies
LS10882H03H25
5 x 5μg Each for $249.00
 
10882H03H5 View Documents Life Technologies
LS10882H03H5
5μg Each for $126.00
 
Description

Description

Ephrin-A1 (EFNA1), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse EFNA2 at 2μg/mL (100μL/well) can bind human EFNA1 with a linear range of 0.8-20ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
Lyophilized
1942
<1 EU/μg
EFNA1, B61, ECKLG, EFL1, EPLG1, LERK1, TNFAIP4

For Research Use Only. Not for use in diagnostic procedures.