Please login to your online account to display your discounted pricing

Sino Biological™ EFNA1 (EphrinA1) Recombinant Human Protein, hIgG1-Fc.His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the mature form of human EFNA1 (NP_004419.2), residues 1 to 182, fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS10882H03H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10882H03H5

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 47.5kDa
Quantity 5μg
Protein Family Kinases & Inhibitors
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Gene Alias EFNA1, B61, ECKLG, EFL1, EPLG1, LERK1, TNFAIP4
Validated Application Assay Standard
Gene ID (Entrez) 1942
Endotoxin Concentration <1 EU/μg
Gene Symbol EFNA1
Protein Subtype Ephrins
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Recombinant/Cell Surface Protein/Ligand
Common Name Ephrin-A1
Protein Tag His Tag/IgG1 Fc Tag
Expression System Mammalian
Format Lyophilized
Purity ≥85% by SDS-PAGE

Ephrin-A1 (EFNA1), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse EFNA2 at 2μg/mL (100μL/well) can bind human EFNA1 with a linear range of 0.8-20ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.