missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ EFNA1 (EphrinA1) Recombinant Human Protein, hIgG1-Fc.His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human EFNA1 (NP_004419.2), residues 1 to 182, fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Manufacturer:  Life Technologies LS10882H03H250

 View more versions of this product

Catalog No. 10882H03HL


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; EFNA1 (EphrinA1) Recombinant Human Protein, hIgG1-Fc.His Tag; 5 x 50ug; Source: Human; Purity: greater than or equal to 85% by SDS-PAGE

Ephrin-A1 (EFNA1), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse EFNA2 at 2μg/mL (100μL/well) can bind human EFNA1 with a linear range of 0.8-20ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
1942
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Ephrins
EFNA1, B61, ECKLG, EFL1, EPLG1, LERK1, TNFAIP4
EFNA1
Kinases & Inhibitors
His Tag/IgG1 Fc Tag
Lyophilized
47.5kDa
5 x 50μg
Recombinant/Cell Surface Protein/Ligand
<1 EU/μg
Ephrin-A1
Assay Standard
Mammalian