missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ EDAR Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human EDAR (NP_071731.1) extracellular domain (Met 1-Ile 189) fused to a polyhistidine tag at the C-terminus

$98.25 - $98.25

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias DL, ED1R, ED3, ED5, EDA-A1R, EDA1R, EDA3, FLJ94390, HRM1
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
11577H08H5 Life Technologies
LS11577H08H5
5μg Each for $98.25
Specifications

Description

Ectodysplasin A Receptor (EDAR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
DL, ED1R, ED3, ED5, EDA-A1R, EDA1R, EDA3, FLJ94390, HRM1
PAGE
Lyophilized
10913