Please login to your online account to display your discounted pricing

Sino Biological™ ECE-1 Recombinant Human Protein, N-His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human ECE1 isoform B (P42892-1) (Gln 90-Trp 770) fused to a polyhistidine tag at the N-terminus

Manufacturer: life technologies  LS10887H07H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10887H07H50

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 80kDa
Purification Method PAGE
Purity ≥90% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Termination N-Terminal
Structural Form Truncated/Recombinant
Endotoxin Concentration <1 EU/μg
Gene Alias RP3-329E20.1, ECE
Common Name Endothelin converting enzyme 1
Gene Symbol ECE1
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Proteases, Peptidases & Inhibitors
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 1889

Endothelin Converting Enzyme 1 (ECE1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.