missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ DUSP3 Recombinant Human Protein, N-His.GST Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human VHR (P51452) (Met 1-Pro 185) fused to the N-terminal polyhistidine-tagged GST tag at the N-terminus

$212.00 - $212.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias DUSP3
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10114H20B25 Life Technologies
LS10114H20B25
5 x 5μg Each for $212.00
Specifications

Description

Dual Specificity Phosphatase 3 (DUSP3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 20mM Tris, 500mM NaCl, 2mM GSH, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
DUSP3
PAGE
Lyophilized
1845