Please login to your online account to display your discounted pricing

Sino Biological™ DUSP3 Recombinant Human Protein, N-His.GST Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human VHR (P51452) (Met 1-Pro 185) fused to the N-terminal polyhistidine-tagged GST tag at the N-terminus

Manufacturer: life technologies  LS10114H20B25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10114H20B25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 48.3kDa
Purification Method PAGE
Purity ≥90% by SDS-PAGE
Quantity 5 x 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Termination N-Terminal
Structural Form Full Length/Recombinant
Endotoxin Concentration <1 EU/μg
Gene Alias DUSP3
Common Name Dual specificity phosphatase 3
Gene Symbol DUSP3
Protein Tag GST Tag/His Tag
Validated Application Assay Standard
Protein Family Other Proteins
Expression System Insect
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 1845

Dual Specificity Phosphatase 3 (DUSP3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 20mM Tris, 500mM NaCl, 2mM GSH, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.