Please login to your online account to display your discounted pricing

Sino Biological™ DPP4 Recombinant Human Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Asn 29-Pro 766) of the mature form of human DPP4 (NP_001926.2) fused to the fused Fc region of human IgG1 at the N-terminus

$227.00 - $227.00

Specifications

Purity ≥95% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias DPP4, ADABP, ADCP2, CD26, DPPIV, TP103
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10688H01H5

DFS Item
 
life technologies
LS10688H01H5
5μg Each for $227.00
Description & Specifications

Specifications

Purity ≥95% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias DPP4, ADABP, ADCP2, CD26, DPPIV, TP103

Dipeptidyl-Peptidase 4 (DPP4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: 1. Measured by its ability to bind recombinant Cynomolgus CXCL12 in a functional ELISA; 2.Measured by its ability to bind recombinant Human SDF1b in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.