missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ DOT1L Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human DOT1L (NP_115871.1) N-terminal segment (Gly 2-Lys 416) fused to two additional amino acids (Gly & Pro) at the N-terminus

$145.00 - $1,744.00

Specifications

Purity ≥85% by SDS-PAGE
Purification Method PAGE
Format Frozen
Gene Alias DKFZp586P1823, DOT1, KIAA1814, KMT4
Gene ID (Entrez) 84444
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11552HNCE5 Life Technologies
LS11552HNCE5
5μg Each for $145.00
11552HNCEL Life Technologies
LS11552HNCE250
5 x 50μg Each for $1,744.00
Specifications

Description

DOT1-like histone H3 methyltransferase (S. cerevisiae) (DOT1L), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM HEPES, 150mM NaCl, 1mM EDTA, 15% glycerol, pH 7.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥85% by SDS-PAGE
Frozen
84444
PAGE
DKFZp586P1823, DOT1, KIAA1814, KMT4