Please login to your online account to display your discounted pricing

Sino Biological™ DOT1L Recombinant Human Protein DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human DOT1L (NP_115871.1) N-terminal segment (Gly 2-Lys 416) fused to two additional amino acids (Gly & Pro) at the N-terminus

Manufacturer: life technologies  LS11552HNCE250

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11552HNCEL

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 47.6kDa
Storage Requirements Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Common Name Dot1-Like, Histone H3 Methyltransferase (S. cerevisiae)
Expression System E. coli
Format Frozen
Quantity 5 x 50μg
Gene Alias DKFZp586P1823, DOT1, KIAA1814, KMT4
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Purification Method PAGE
Protein Family Methylation & Demethylation Proteins
Structural Form Truncated/Recombinant
Validated Application Assay Standard
Purity ≥85% by SDS-PAGE
Gene Symbol DOT1L
Protein Tag Untagged
Gene ID (Entrez) 84444

DOT1-like histone H3 methyltransferase (S. cerevisiae) (DOT1L), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM HEPES, 150mM NaCl, 1mM EDTA, 15% glycerol, pH 7.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.