missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DMP1 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human DMP1 (Q13316-1) (Met 1-Tyr 513) fused to a polyhistidine tag at the C-terminus

$366.00 - $366.00

Specifications

Purity >85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias ARHP, ARHR, DMP-1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11929H08H50 View Documents Life Technologies
LS11929H08H50
50μg Each for $366.00
 
Description

Description

Dentin Matrix Acidic Phosphoprotein 1 (DMP1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind human CFH in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

>85% by SDS-PAGE
<1 EU/μg
Lyophilized
ARHP, ARHR, DMP-1

For Research Use Only. Not for use in diagnostic procedures.