missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ DMBT1 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the N-terminal segment of human DMBT1 (NP_004397.2) (Met 1-Ser 220) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Life Technologies LS11678H08H250

 View more versions of this product

Catalog No. 11678H08HL


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; DMBT1 Recombinant Human Protein, His Tag; 5 x 50ug; Source: Human; Purity: greater than or equal to 95% by SDS-PAGE

Deleted In Malignant Brain Tumors 1 (DMBT1) (GP340), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human Galectin-3 (Mfr. No. 10289-HNAE-E) at 10μg/mL (100μL/well) can bind DMBT1 with a linear range of 0.06-1.0μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
22.6kDa
5 x 50μg
Other Proteins|Glycoproteins
DMBT1, RP11-481L19.1, FLJ61058, GP340, mgC164738, muclin
DMBT1
Other Proteins
His Tag
Lyophilized
≥95% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Deleted In Malignant Brain Tumors 1 ( DMBT1 / GP340 )
Assay Standard
Mammalian