missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DDC Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the full length of human DDC (NP_000781.1) (Met 1-Glu 480) fused to a polyhistidine tag at the C-terminus

$644.00 - $644.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Frozen
Gene Alias AADC, DDC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10560H08B25 View Documents Life Technologies
LS10560H08B25
5 x 5μg Each for $644.00
 
Description

Description

Dopa Decarboxylase (DDC), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to convert the substrate 3,4-dihydroxy L-phenylalanine (L-Dopa) to 3,4-dihydroxyphenylethylamine (dopamine); the dopamine product is measured by its absorbance at 340nM after derivatization with trinitrobenzene sulfonic acid, the specific activity is >1500pmole/min./μg
  • Formulation: 50mM Tris, 100mM NaCl, 0.2μg/μL BSA, pH 8, 10% glycerol
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
<1 EU/μg
Frozen
AADC, DDC

For Research Use Only. Not for use in diagnostic procedures.