missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ DDC Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the full length of human DDC (NP_000781.1) (Met 1-Glu 480) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Life Technologies LS10560H08B5

 View more versions of this product

Catalog No. 10560H08B5


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; DDC Recombinant Human Protein, His Tag; 5ug; Source: Human; Purity: greater than or equal to 85% by SDS-PAGE

Dopa Decarboxylase (DDC), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to convert the substrate 3,4-dihydroxy L-phenylalanine (L-Dopa) to 3,4-dihydroxyphenylethylamine (dopamine); the dopamine product is measured by its absorbance at 340nM after derivatization with trinitrobenzene sulfonic acid, the specific activity is >1500pmole/min./μg
  • Formulation: 50mM Tris, 100mM NaCl, 0.2μg/μL BSA, pH 8, 10% glycerol
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
55kDa
5μg
Other Proteins
AADC, DDC
DDC
Insect
Frozen
≥85% by SDS-PAGE
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Dopa Decarboxylase
Assay Standard
His Tag