missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CXCL9 Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human CXCL9 (Q07325) (Thr 23-Thr 125)

$313.00 - $313.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CMK, Humig, MIG, SCYB9, crg-10
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10888HNAE25 Life Technologies
LS10888HNAE25
5 x 5μg Each for $313.00
Specifications

Description

Chemokine (C-X-C Motif) Ligand 9 (CXCL9), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
CMK, Humig, MIG, SCYB9, crg-10
PAGE
Lyophilized
4283