Please login to your online account to display your discounted pricing

Sino Biological™ CXCL9 Recombinant Human Protein DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human CXCL9 (Q07325) (Thr 23-Thr 125)

Manufacturer: life technologies  LS10888HNAE25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10888HNAE25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 11.9kDa
Purification Method PAGE
Purity ≥90% by SDS-PAGE
Quantity 5 x 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Structural Form Full Length/Recombinant
Protein Subtype Other Cytokines
Endotoxin Concentration <1 EU/μg
Gene Alias CMK, Humig, MIG, SCYB9, crg-10
Common Name Chemokine (C-X-C motif) ligand 9
Gene Symbol CXCL9
Validated Application Assay Standard
Protein Family Chemokines & Receptors
Expression System E. coli
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 4283

Chemokine (C-X-C Motif) Ligand 9 (CXCL9), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.