missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ CXCL2 Recombinant Mouse Protein, N-His.SUMO Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of mouse CXCL2 (P10889-1) (Ala 28-Asn 100) fused to a N-terminal polyhistide-tagged SUMO tag at the N-terminus

$313.00 - $313.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CINC-2a, GROb, Gro2, MIP-2, MIP-2a, mgsa-b, Mip2, Scyb, Scyb2
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of protein in suspension buffer


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50070M28E25 Life Technologies
LS50070M28E25
5 x 5μg Each for $313.00
Specifications

Description

Chemokine (C-X-C Motif) Ligand 2 (CXCL2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 0.1% TFA, pH 2.9, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
CINC-2a, GROb, Gro2, MIP-2, MIP-2a, mgsa-b, Mip2, Scyb, Scyb2
PAGE
Lyophilized
20310