missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CXCL16 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mouse CXCL16 (NP_075647.3) extracellular domain (Met 1-Trp 201) fused to a C-terminal polyhistidine tag

$280.00 - $280.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias DN-491F10.6, 0910001K24Rik, AV290116, BB024863, SR-PSOX, Zmynd15, CXCL16
View More Specs

Includes: Five vials of protein in suspension buffer

Products 1
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
50142M08H25 missing translation for 'viewDocuments' Life Technologies
LS50142M08H25
5 x 5μg Each for $280.00
 
Description

Description

Chemokine (C-X-C Motif) Ligand 16 (CXCL16), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
DN-491F10.6, 0910001K24Rik, AV290116, BB024863, SR-PSOX, Zmynd15, CXCL16
PAGE
Lyophilized
66102

For Research Use Only. Not for use in diagnostic procedures.